Learn More
BioVision IncSupplier Diversity Partner Mitochondrial DNA Isolation Kit: Mitochondrial DNA Isolation Kit: Rapid & Convenient Kit for Isolating mtDNA from a variety of Cells and Tissues. High yield and Purity. No genomic DNA contamination. 50 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K28050
Mitochondria are semiautonomous organelles which functions in aging process, apoptosis, anti-HIV drugs, and cancers. Mitochondrial DNA (mtDNA) has a very high mutation rate and the mutations on mtDNA appear to be related to certain diseases such as diabetes, Alzheimer’s disease, and muscle disorders. Isolation and quantification of mtDNA are often required to study the relationships between the diseases and mtDNA. The Mitochondrial DNA Extraction Kit provides convenient tools for isolating mtDNA from a variety of cells and tissues in high yield and purity, without contaminations from genomic DNA. The purified mtDNA can be used for a variety of studies such as enzyme manipulations, Southern blotting, cloning, PCR analysis, and amplifications. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9664