Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Mitochondria Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mitochondria Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS1372P0

 View more versions of this product

Catalog No. MS1372P0


Add to cart

Description

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mitochondria Ab-2, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MTC02

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified mitochondrial preparation

Molecular Weight: 60kDa

Positive Control: HeLa cells, Liver

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Mitochondria Ab-2
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG1
RUO
Primary
Monoclonal
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
MTC02
Unconjugated
Concentrated
Semi-purified mitochondrial preparation
100μL
Cell Cycle
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit