Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Thermo Scientific™ Lab Vision™ Mitochondria Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mitochondria Ab-2, Mouse Monoclonal Antibody.

$242.50 - $923.00

Specifications

Antigen Mitochondria Ab-2
Clone MTC02
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1372B
View Documents
Thermo Scientific™
MS1372B
Biotinylated Ab with BSA and Azide 1mL Each for $923.00
N/A
MS1372B0
View Documents
Thermo Scientific™
MS1372B0
Biotinylated Ab with BSA and Azide 100μL Each for $242.50
N/A
MS1372B1
View Documents
Thermo Scientific™
MS1372B1
Biotinylated Ab with BSA and Azide 500μL Each for $596.00
N/A
MS1372P
View Documents
Thermo Scientific™
MS1372P
Purified Ab with BSA and Azide 1mL Each for $897.00
N/A
MS1372P0
View Documents
Thermo Scientific™
MS1372P0
Purified Ab with BSA and Azide 100μL Each for $248.00
N/A
MS1372P1
View Documents
Thermo Scientific™
MS1372P1
Purified Ab with BSA and Azide 500μL Each for $614.50
N/A
MS1372P1ABX
View Documents
Thermo Scientific™
MS1372P1ABX
Ab Without BSA/Azide (Frozen) 100μL Each for $614.00
N/A
MS1372PABX
View Documents
Thermo Scientific™
MS1372PABX
Ab Without BSA/Azide (Frozen) 200μL Each for $729.00
N/A
MS1372R7
View Documents
Thermo Scientific™
MS1372R7
Ready-to-use for IHC (UVLP) 7mL Each for $393.00
N/A
MS1372R1
View Documents
Thermo Scientific™
MS1372R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Mitochondria Ab-2, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MTC02

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified mitochondrial preparation

Molecular Weight: 60kDa

Positive Control: HeLa cells, Liver

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Mitochondria Ab-2
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Murine
Cell Cycle
Semi-purified mitochondrial preparation
MTC02
Monoclonal
RUO
Primary
IgG1
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit