Learn More
Mitochondria Mouse anti-Human, Clone: AE-1, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB14641
Catalog No. 89-034-724
Add to cart
Description
Description
Mouse monoclonal antibody raised against human mitochondria.
  • High specificity
  • High reactivity
Specifications
Specifications
Mitochondria
AE-1
Mouse monoclonal antibody raised against human mitochondria.
In 10 mM PBS (0.05% BSA and 0.05% azide).
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot ( 0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Semi-purified mitochondrial preparation.
Protein A/G purification
RUO
Primary
This antibody recognizes a 60 kDa antigen associated with the mitochondria in human cells. It reacts specifically with human cells, including neurons and embryonic stem cells.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)