Learn More
BioVision IncSupplier Diversity Partner Mitochondrial Protein IP Kit: Mitochondrial Protein IP Kit: Ready-to-use reagents for IP/Co-IP/Pull down of mt. proteins or complexes. Compatible with SDS PAGE, 2D gel, Blue-Native gel, Mass spectrometry. 50 assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K28550
Mitochondrial Protein IP Kit: Mitochondria are the power house of the cells and play an essential role in energy production. Mitochondrial dysfunction leads to several disorders like cardiac dysfunction, diabetes, aging and neurological disorder. BioVision ready to use mitochondria Protein IP Buffer is optimized for immunoprecipitation (IP and co-IP) using mitochondria and mitochondrial extracts. The buffer is a gentle formulation, which maintains the stability of mitochondrial complexes. The Mitochondrial Protein IP kit is provided with different choices of detergents like n-Dodecyl-beta-D-maltoside, Triton X-100 and digitonin to achieve different stringency conditions for protein-protein interaction studies. Triton X-100 is the most commonly used detergent especially for membrane protein solubilization. However, in case of fragile complexes digitonin or n-Dodecyl-beta-D-maltoside is the choice of detergents. BioVision charges an Ice pack fee for each order.
Catalog No. 50-111-9666