Learn More
Mitofusin 2 Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: S153-5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242188UV
Catalog No. NBP242188UV
Add to cart
Description
Description
Mitofusin 2 Monoclonal antibody specifically detects Mitofusin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Mitofusin 2
S153-5
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
MFN2
Fusion protein amino acids 370-600 (cytoplasmic N-terminus) of mouse Mitofusin-2. Rat: 97% identity (226/231 amino acids identical). Human: 92% identity (214/231 amino acids identical) ~55% identity with Mitofusin-1.
Protein G purified
Store at 4C in the dark.
Monoclonal
Approx 55% identity with Mitofusin-1.Detects approx 90kDa. No cross-reactivity against Mitofusin-1.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CMT2A, CMT2A2, CPRP1mitochondrial assembly regulatory factor, EC 3.6.5, EC 3.6.5.-, HSG, KIAA0214hyperplasia suppressor, MARF, mitofusin 2, mitofusin-2, Transmembrane GTPase MFN2
Mouse
IgG2a
0.1 ml
Primary
9927
Human, Mouse, Rat