Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mitofusin2 Rabbit anti-Human, DyLight 488, Polyclonal,Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP266383G
Description
Mitofusin2 Polyclonal antibody specifically detects Mitofusin2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Mitofusin2 | |
DyLight 488 | |
Affinity Purified | |
CMT2A, CMT2A2, CPRP1mitochondrial assembly regulatory factor, EC 3.6.5, EC 3.6.5.-, HSG, KIAA0214hyperplasia suppressor, MARF, mitofusin 2, mitofusin-2, Transmembrane GTPase MFN2 | |
MFN2 | |
Partial recombinant human MFN2 protein (amino acids 364-599). [O95140] | |
Polyclonal | |
Immunogen affinity purified | |
Mitochondrial Markers | |
Human,Mouse,Rat |
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
50mM Sodium Borate with 0.05% Sodium Azide | |
9927 | |
Rabbit | |
IgG | |
Primary | |
0.1mL | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title