Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mitomycin C

Click to view available options
Quantity:
5mg
10mg
50-07-7
C15H18N4O5
334.332
MFCD00078109
NWIBSHFKIJFRCO-WUDYKRTCSA-N
5746
CHEBI:27504
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N
50-07-7
C15H18N4O5
334.332
MFCD00078109
NWIBSHFKIJFRCO-WUDYKRTCSA-N
5746
CHEBI:27504
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N
Solubility
Soluble in methanol, water and dimethyl sulfoxide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong bases and strong acids.
UN Number UN2811
Beilstein 7231816
Merck Index 14,6215
Solubility Information Soluble in methanol at 10mg/ml; Also soluble in water at 1mg/ml with warming; Soluble in DMSO at ≈15mg/ml
Formula Weight 334.33
Physical Form Powder
Color Blue to Purple
Melting Point 360°C
Quantity 5mg
Chemical Name or Material Mitomycin C

GHS H Statement
H300-H351
Fatal if swallowed.
Suspected of causing cancer.

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CN0700000

TSCA : TSCA

Recommended Storage : 2° to 8°C