Learn More
Thermo Scientific™ MitoSensor Orange Dye
Label live mitochondria with orange fluorescence.
Specifications
Description Cellomics MitoSensor Orange
View More Specs
Products 1
Catalog Number Mfr. No. Description Quantity Target Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Target Price Quantity & Availability  
84-096-01
View Documents
Thermo Scientific™
8409601
Cellomics MitoSensor Orange 5 x 20µg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Label live mitochondria with orange fluorescence using Thermo Scientific™ MitoSensor Orange Dye.
The selective labeling of live cell compartments provides a powerful method for studying cellular events in a spatial and temporal context.
Proprietary Dye
  • Selectively accumulates in mitochondria when the mitochondrial membrane potential gradient is not compromised
  • Mitochondrial indicator is a hydrophobic compound that easily penetrates intact live cells, and labels mitochondria after it enters the cells
  • Fluorescent mitochondrial indicator is retained in mitochondria for long time since the indicator carries a cell-retaining group
  • Mitochondrial dye can be fixed in the cell, significantly increasing the staining efficiency of this product
Robust labeling protocol requires minimal hands-on time
  • Protocol can be readily adapted for a wide variety of fluorescence platforms, such as fluorescence microscopy, microplate assays, immunocytochemistry and flow cytometry
  • Useful for a variety of studies, including cell adhesion, chemotaxis, multidrug resistance, cell viability, apoptosis and cytotoxicity
  • Suitable for proliferating and non-proliferating cells; can be used for both suspension and adherent cells
Specifications
Specifications
Cellomics MitoSensor Orange