Learn More
Mitosis and Meiosis Lab Investigations
An exciting and interactive way to learn about mitosis and meiosis.
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
S32617
![]() |
Neo/Sci Corporation
201283 |
Students learn about the formation of sex cells | Meiosis Simulation Lab Investigation Kit |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Construct models using colored pop beads to illustrate chromosome structure and function.
Models can also be used to depict:
- Processes that trigger cell division
- Distinction between plant and animal mitosis
- Crossing over
- Genetic recombination
- Chromosomal aberration
Order Info
Save money when you buy these kits with their corresponding CD-ROM as a curriculum pack.
Specifications
40 students |