Learn More
Selleck Chemical LLCSupplier Diversity Partner Mitoxantrone 2HCl 100mg 70476-82-3 NSC-301739  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S2485100MG
Mitoxantrone 2HCl (NSC-301739) is a dihydrochloride salt of Mitoxantrone. Mitoxantrone is an inhibitor of type II topoisomerase and protein kinase C (PKC) with IC50 of 8.5 μM for PKC. Mitoxantrone inhibits cell proliferative growth of MCF-7/wt cells with IC50 of 0.42 μM. Mitoxantrone also induces apoptosis.
Catalog No. 50-698-2