Learn More
Jaece Mity Plugs Cellulose Acetate Plugs
Plug drosophila vials and bottles. Mity Plugs Cellulose Acetate Plugs have a non-shedding, dense weave that offers a superior barrier to mite infestation. They allow reduced food evaporation to keep food fresh for longer.
Manufacturer:  Jaece JZG710
 View more versions of this product
Catalog No. 14-127-105
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Fits 24mm Glass Vials
1000/Cs.
Cellulose Acetate
Mity Drosophila Plug