Learn More
TARGETMOL CHEMICALS INC Djenkolic Acid 25MG
Supplier: TARGETMOL CHEMICALS INC T2231125MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Djenkolic Acid (S,S'-Methylenebiscysteine) is a naturally occurring sulfur-containing amino acid extracted from djenkol beans(Archidendron pauciflorum), which is native to Southeast Asia. It is an uncommon but important cause of acute kidney injury in human. Purity 98.77%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.