Learn More
Mixed Calibration Standard 5 For Environmental Analysis, SPEX CertiPrep
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$182.00 - $305.00
Specifications
Chemical Name or Material | Mixed Calibration Standard 5 |
---|---|
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation |
Grade | Environmental |
Solvent or Matrix | 5% HNO3/tr Tart. Acid/tr HF |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
MIXSTD5-100
|
SPEX CertiPrep![]() MIXSTD5100 |
125mL | Triple-leached LDPE bottle |
Each for $182.00
|
|
MIXSTD5-500
|
SPEX CertiPrep![]() MIXSTD5500 |
500mL | Triple-leached LDPE bottle |
Each for $305.00
|
|
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Mg | 1000µg/mL |
Sb | 200µg/mL |
Tl | 200µg/mL |
B | 100µg/mL |
Ag | 50µg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Mixed Calibration Standard 5 | |
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation | |
Environmental | |
5% HNO3/tr Tart. Acid/tr HF |
Safety and Handling
Causes severe skin burns and eye damage
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.