Learn More
Mixed Reagent Solution for Aluminum, Reagents
Mixed Reagent Solution for Aluminum, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CM0100001A
Specifications
| Mixed Reagent Solution for Aluminum | |
| 7732-18-5 , 10034-99-8 , 50-81-7 | |
| Reagent | |
| 1.1226 | |
| Non-sterile |
| Colorless | |
| 102°C | |
| Poly Bottle | |
| 1 L | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.