Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Mixter Right Angle Forceps, Sklar 9-1/4 in., Right Angled (90 Degrees), Serrated, Heavy, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 552892
Forceps, Mixter Right Angle Forceps, Sklar, 9-1/4 in., Right Angled (90 Degrees), Serrated, Heavy, Stainless Steel, EA. Sklar Mixter Right Angle Forceps are ratcheted, finger ring forceps commonly used for working in obscured surgical sites. They are most frequently used for clamping, dissection, or grasping tissue. Mixter Right Angle Forceps are available in multiple lengths and have serrations the entire length of the jaw. This product has straight shanks, heavy, fully serrated jaws, and right angle tips. The length is 9-1/4 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-111-8908