Learn More
Azzota Corp MK-1775 DFS Item
Manufacturer:  Azzota Corp C1223
MK-1775 (AZD-1775) is a WEE1 inhibitor. MK-1775 selectively targets and inhibits WEE1, a tyrosine kinase that phosphorylates cyclin-dependent kinase 1 (CDC2) to inactivate the CDC2/cyclin B complex. Please request if need more quantities
Catalog No. 50-161-3148