Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MK 1903, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective hydroxycarboxylic acid receptor 2 (HCA2, GPR109A) full agonist; exhibits greater potency than niacin in a whole cell HTRF-cAMP assay (EC50 values are 12.9 and 51 nM respectively). Exhibits no binding at the GRP109B receptor.
Specifications
Color | White |
Quantity | 50 mg |
Synonym | unii-62n05gri0p, 1ar, 5ar-1a,3,5,5a-tetrahydro-1h-2,3-diaza-cyclopropa a pentalene-4-carboxylic acid, 1ar,5ar-1a,2,5,5a-tetrahydro-1h-2,3-diazacyclopropa a pentalene-4-carboxylic acid, 1ar,5ar-1a,3,5,5a-tetrahydro-1h-2,3-diaza-cyclopropa a pentalene-4-carboxylic acid, 2r,4r-8,9-diazatricyclo 4.3.0.0 2 ,? nona-1 6 ,7-diene-7-carboxylic acid, 4ar,5ar-4,4a,5,5a-tetrahydro-1h-cyclopropa 4,5 cyclopenta 1,2-c pyrazole-3-carboxylic acid, 4ar,5ar-4,4a,5,5a-tetrahydro-1h-cyclopropa 4,5 cyclopenta 1,2 pyrazole-3-carboxylic acid, 1h-cyclopropa 4,5 cyclopenta 1,2-c pyrazole-3-carboxylic acid, 4,4a,5,5a-tetrahydro-, 4ar,5ar |
Formula Weight | 164.16 |
Percent Purity | >98% |
Physical Form | Solid |
Chemical Name or Material | MK 1903 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction