Learn More
Selleck Chemical LLCSupplier Diversity Partner Ostarine (GTx-024) 50mg 841205-47-8 MK-2866  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S117450MG
Ostarine (GTx-024, MK-2866) is a selective androgen receptor modulator (SARM) with Ki of 3.8 nM, and is tissue-selective for anabolic organs. Phase 3.
Catalog No. 50-689-4