Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-MK 801 maleate, Tocris Bioscience™

Catalog Number 095510
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
121917-57-5
C20H19NO4
337.375
QLTXKCWMEZIHBJ-FWHYOZOBSA-N
--mk 801 maleate, unii-ef13o02iv0, dizocilpine maleate, --mk-801 hydrogen maleate, dizocilpine maleate,-, -mk-801 hydrogen maleate, --mk 801 hydrogen maleate, --mk-801 hydrogen maleate hplc, 5r,10s---5-methyl-10,11-dihydro-5h-dibenzo a,d cylcohepten-5,10-imine maleate
16219612
CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O
This item is not returnable. View return policy
121917-57-5
C20H19NO4
337.375
QLTXKCWMEZIHBJ-FWHYOZOBSA-N
--mk 801 maleate, unii-ef13o02iv0, dizocilpine maleate, --mk-801 hydrogen maleate, dizocilpine maleate,-, -mk-801 hydrogen maleate, --mk 801 hydrogen maleate, --mk-801 hydrogen maleate hplc, 5r,10s---5-methyl-10,11-dihydro-5h-dibenzo a,d cylcohepten-5,10-imine maleate
16219612
CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O

NMDA antagonist; less active enantiomer of (+)-MK 801

Less active enantiomer (10-fold less potent in behavioral studies) than (+)-MK 801 maleate.

(+)-enantiomer also available.

Quantity 10 mg
Formula Weight 337.37
Percent Purity >99%
Chemical Name or Material (-)-MK 801 maleate
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit