Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mKate Mouse anti-Tag, Magnetic beads, Clone: OTI1G5, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA183041

Description
mKate fluorescent protein.Specifications
mKate | |
Monoclonal | |
PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide | |
Mouse | |
Affinity Chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Immunoprecipitation | |
OTI1G5 | |
mKate | |
Recombit protein and recognizes mKate. | |
1 mL | |
Primary | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction