Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKI67 Mouse anti-Human, Unconjugated, Clone: MKI67/2461, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB21292
Description
MKI67 Monoclonal antibody specifically detects MKI67 in Human samples. It is validated for IHC-P, IF, Flow Cyt.Specifications
MKI67 | |
MKI67/2461 | |
antigen identified by monoclonal antibody Ki-67 | |
Liquid | |
Mouse | |
IgG1, Kappa | |
Store at 2 to 8°C | |
Monoclonal | |
Human |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
MKI67 | |
KIA, Ki-67 | |
Recombinant protein corresponding to amino acids 2293-2478 of human MKI67 | |
100 μg | |
Primary | |
4288 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title