Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKK3/MEK3 Rabbit anti-Human, Clone: SD20-93, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP267747

Catalog No. NBP267747


Add to Cart

Description

Description

MKK3/MEK3 Monoclonal specifically detects MKK3/MEK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

MKK3/MEK3
Monoclonal
1 mg/mL
Western Blot 1:1000-1:5000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200
MAP kinase kinase 3, MAPK/ERK kinase 3, MAPKK 3, MAPKK3PRKMK3dual specificity mitogen-activated protein kinase kinase 3, MEK3MEK 3, mitogen activated protein kinase kinase 3, mitogen-activated protein kinase kinase 3, MKK3EC 2.7.12.2
Rabbit
Protein A purified
Angiogenesis, Cancer, Hypoxia, Lipid and Metabolism, Phospho Specific, Protein Kinase
5606
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
SD20-93
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
MAP2K3
Recombinant protein within human MKK3/MEK3 aa 101-347/347. (SwissProt: P46734 Human; SwissProt: O09110 Mouse; Unigene: 100064 Rat)
100 μL
Primary
Human, Mouse, Rat
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit