Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKK7/MEK7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15500120UL

 View more versions of this product

Catalog No. NBP1550020


Add to cart

Description

Description

MKK7/MEK7 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

MKK7/MEK7
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
MAP2K7
Synthetic peptides corresponding to MAP2K7(mitogen-activated protein kinase kinase 7) The peptide sequence was selected from the middle region of MAP2K7. Peptide sequence ERIDPPDPTKPDYDIRADVWSLGISLVELATGQFPYKNCKTDFEVLTKVL.
47 kDa
20ul
Protein Kinase
Primary
5609
Western Blot
Western Blot 1:100-1:2000
O14733
c-Jun N-terminal kinase kinase 2, dual specificity mitogen-activated protein kinase kinase 7, EC 2.7.12.2, JNK kinase 2, JNK-activating kinase 2, JNKK 2, JNKK2, MAP kinase kinase 7, MAPK/ERK kinase 7, MAPKK7, MEK 7, MEK7, mitogen-activated protein kinase kinase 7, MKK7MAPKK 7, PRKMK7
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit