Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKL1 Mouse anti-Human, Clone: OTI4F6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene CF805132
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene interacts with the transcription factor myocardin, a key regulator of smooth muscle cell differentiation. The encoded protein is predomitly nuclear and may help transduce signals from the cytoskeleton to the nucleus. This gene is involved in a specific translocation event that creates a fusion of this gene and the RNA-binding motif protein-15 gene. This translocation has been associated with acute megakaryocytic leukemia.Specifications
MKL1 | |
Monoclonal | |
Unconjugated | |
MRTFA | |
BSAC, MAL, MKL, MRTF-A | |
Mouse | |
Affinity Chromatography | |
RUO | |
57591 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4F6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q969V6 | |
MRTFA | |
Human recombit protein fragment corresponding to amino acids 165-397 of human MKL1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction