Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

MKRN1 Mouse anti-Human, Clone: OTI2C8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525817

Catalog No. PIMA525817



Description

Description

The Makorin ring finger protein-1 gene (MKRN1) is a highly transcribed, intron-containing source for a family of intronless mammalian genes encoding a novel class of zinc finger proteins. Phylogenetic analyses indicate that the MKRN1 gene is the ancestral founder of this gene family (Gray et al., 2000 [PubMed 10843807]).
Specifications

Specifications

MKRN1
Monoclonal
0.98 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHC7
RNF61, E3 ubiquitin-protein ligase makorin-1, RING finger protein 61
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23608
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2C8
Unconjugated
MKRN1
Liquid
MKRN1
Full length human recombit protein of MKRN1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit