Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKRN3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281964MFV500
Description
MKRN3 Polyclonal antibody specifically detects MKRN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MKRN3 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| EC 6.3.2.-, makorin ring finger protein 3, MGC88288, RNF63D15S9, ZFP127RING finger protein 63, Zinc finger protein 127, ZNF127probable E3 ubiquitin-protein ligase makorin-3 | |
| MKRN3 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MKRN3. The immunogen is located within amino acids 400 - 450 of MKRN3. Amino Acid Squence: FYKHEYPEGWGDEPPG | |
| 0.1 mL | |
| Zinc Finger | |
| 7681 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction