Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 161, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
423735-93-7
C17H17BrN2O2
361.239
DFOVLSMXPWPCFH-UHFFFAOYSA-N
2-bromo-n-3-butyramidophenyl benzamide,2-bromo-n-3-butanoylamino phenyl benzamide,2-bromo-n-3-1-oxobutyl amino phenyl benzamide,2-bromo-n-3-butyrylamino phenyl benzamide,cambridge id 5946339,aronis27179,benzamide, 2-bromo-n-3-1-oxobutyl amino phenyl,ml hplc,n-3-butyrylamino phenyl-2-bromobenzamide,2-bromo-n-3-butanamidophenyl benzamide
1048267
2-bromo-N-[3-(butanoylamino)phenyl]benzamide
CCCC(=O)NC1=CC=CC(=C1)NC(=O)C2=CC=CC=C2Br
This item is not returnable. View return policy
423735-93-7
C17H17BrN2O2
361.239
DFOVLSMXPWPCFH-UHFFFAOYSA-N
2-bromo-n-3-butyramidophenyl benzamide,2-bromo-n-3-butanoylamino phenyl benzamide,2-bromo-n-3-1-oxobutyl amino phenyl benzamide,2-bromo-n-3-butyrylamino phenyl benzamide,cambridge id 5946339,aronis27179,benzamide, 2-bromo-n-3-1-oxobutyl amino phenyl,ml hplc,n-3-butyrylamino phenyl-2-bromobenzamide,2-bromo-n-3-butanamidophenyl benzamide
1048267
2-bromo-N-[3-(butanoylamino)phenyl]benzamide
CCCC(=O)NC1=CC=CC(=C1)NC(=O)C2=CC=CC=C2Br

Inhibits PAR1-mediated platelet activation

Inhibitor of protease-activated receptor 1 (PAR1)-mediated platelet activation (IC50 = 0.26 μM for the inhibition of platelet P-selectin expression on human platelets). Thought to act allosterically. Also inhibits thrombin-induced platelet activation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 361.23
Percent Purity >98%
Chemical Name or Material ML 161

Recommended Storage : Store at +4°C