Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 161, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
423735-93-7
C17H17BrN2O2
361.239
DFOVLSMXPWPCFH-UHFFFAOYSA-N
2-bromo-n-3-butyramidophenyl benzamide,2-bromo-n-3-butanoylamino phenyl benzamide,2-bromo-n-3-1-oxobutyl amino phenyl benzamide,2-bromo-n-3-butyrylamino phenyl benzamide,cambridge id 5946339,aronis27179,benzamide, 2-bromo-n-3-1-oxobutyl amino phenyl,ml hplc,n-3-butyrylamino phenyl-2-bromobenzamide,2-bromo-n-3-butanamidophenyl benzamide
1048267
2-bromo-N-[3-(butanoylamino)phenyl]benzamide
CCCC(=O)NC1=CC=CC(=C1)NC(=O)C2=CC=CC=C2Br
This item is not returnable. View return policy
423735-93-7
C17H17BrN2O2
361.239
DFOVLSMXPWPCFH-UHFFFAOYSA-N
2-bromo-n-3-butyramidophenyl benzamide,2-bromo-n-3-butanoylamino phenyl benzamide,2-bromo-n-3-1-oxobutyl amino phenyl benzamide,2-bromo-n-3-butyrylamino phenyl benzamide,cambridge id 5946339,aronis27179,benzamide, 2-bromo-n-3-1-oxobutyl amino phenyl,ml hplc,n-3-butyrylamino phenyl-2-bromobenzamide,2-bromo-n-3-butanamidophenyl benzamide
1048267
2-bromo-N-[3-(butanoylamino)phenyl]benzamide
CCCC(=O)NC1=CC=CC(=C1)NC(=O)C2=CC=CC=C2Br

Inhibits PAR1-mediated platelet activation

Inhibitor of protease-activated receptor 1 (PAR1)-mediated platelet activation (IC50 = 0.26 μM for the inhibition of platelet P-selectin expression on human platelets). Thought to act allosterically. Also inhibits thrombin-induced platelet activation.
Quantity 50mg
Formula Weight 361.23
Percent Purity >98%
Chemical Name or Material ML 161

Recommended Storage : Store at +4°C