Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 162

Inhibits glutathione peroxidase (GPX4); induces ferroptosis
$186.40 - $758.50
Specifications
Chemical Name or Material | 2-[(Chloroacetyl)(3-chloro-4-methoxyphenyl)amino]-N-(2-phenylethyl)-2-thien-2-ylacetamide |
---|---|
CAS | 1035072-16-2 |
Target | Glutathione Peroxidase 4/GPX4 |
Molecular Formula | C23H22Cl2N2O3S |
Purity | 0.98 |
Description
ML 162 is a covalent glutathione peroxidase 4 (GPX4) inhibitor. It induces ferroptosis in vivo. It is selectively lethal to mutant RAS oncogene-expressing cell lines (IC50 = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts, respectively).Specifications
2-[(Chloroacetyl)(3-chloro-4-methoxyphenyl)amino]-N-(2-phenylethyl)-2-thien-2-ylacetamide | |
Glutathione Peroxidase 4/GPX4 | |
0.98 |
1035072-16-2 | |
C23H22Cl2N2O3S |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title