Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (±)-ML 209

RORγt inverse agonist; suppresses Th17 cell differentiation

$196.10 - $823.50

Specifications

CAS 1334526-14-5
Formula Weight 441.52g/mol
For Use With (Application) Agonist against RORγt
Molecular Formula C25H31NO6
Solubility Soluble to 100mM in DMSO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
59-871-0 View Documents Tocris Bioscience™
5987/10
10mg Each for $196.10
Add to cart
 
59-875-0 View Documents Tocris Bioscience™
5987/50
50mg Each for $823.50
Add to cart
 
Description

Description

RORγt inverse agonist (IC50 = 460 nM). Suppresses Th17 cell differentiation and IL17A expression in vitro. Does not suppress transcription of RORα. Minimal activity displayed on ERα, LXRα and thyroid hormone receptors (IC50 values >4.4 μM).
Specifications

Specifications

1334526-14-5
Agonist against RORγt
Soluble to 100mM in DMSO
White
441.52g/mol
C25H31NO6
Store at 4°C

For Laboratory Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit