Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 213, Tocris Bioscience™
GREENER_CHOICE

Catalog No. p-4817476
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

KV7.2 and KV7.4 channel opener

Selective KV7.2 (KCNQ2) and KV7.4 (KCNQ4) channel opener (EC50 values are 230 and 510 nM for KV7.2 and KV7.4 respectively). Displays > 80-fold selectivity against KV7.1, KV7.3 and KV7.5 in a thallium-based fluorescence assay. Hyperpolarizes resting membrane potential in an in vitro spinal cord preparation.

Chemical Identifiers

CAS 489402-47-3
Molecular Formula C17H23NO
Molecular Weight (g/mol) 257.377
InChI Key SIQGKPGBLYKQBB-UHFFFAOYSA-N
Synonym n-mesitylbicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl-bicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-3-carboxamide, bicyclo 2.2.1 heptane-2-carboxylic acid 2,4,6-trimethyl-phenyl-amide, d0xy2m, qcr-50, regid_for_cid_3111211, bicyclo 2.2.1 hept-2-yl-n-2,4,6-trimethylphenyl carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-6-carboxamide
PubChem CID 3111211
IUPAC Name N-(2,4,6-trimethylphenyl)bicyclo[2.2.1]heptane-3-carboxamide
SMILES CC1=CC(=C(C(=C1)C)NC(=O)C2CC3CCC2C3)C

Specifications

Quantity 10 mg
Formula Weight 257.37
Percent Purity >99%
Chemical Name or Material ML 213
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.