Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 239

Breast cancer stem cell inhibitor

Manufacturer:  Tocris Bioscience™ 4829/10

 View more versions of this product

Catalog No. 482910

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

Breast cancer stem cell inhibitor (IC50 = 1.16µM). Exhibits 24-fold selectivity for breast cancer stem cells over normal mammary epithelial cells. Also cytotoxic towards MDA-MB-231 breast cancer cells in vitro.
Specifications

Specifications

2-(2,4,6-Trichlorophenoxy)acetic acid (2E)-2-(1H-pyrrol-2-ylmethylene)hydrazide
C13H10Cl3N3O2
10mg
>98%
1378872-36-6
346.6
Target: Stem Cell Signaling Compounds
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit