Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ML 239, Tocris Bioscience
Breast cancer stem cell inhibitor
Manufacturer: Tocris Bioscience 4829/25
This item is not returnable.
View return policy
Description
Breast cancer stem cell inhibitor (IC50 = 1.16 μM). Exhibits 24-fold selectivity for breast cancer stem cells over normal mammary epithelial cells. Also cytotoxic towards MDA-MB-231 breast cancer cells in vitro.Specifications
2-(2,4,6-Trichlorophenoxy)acetic acid (2E)-2-(1H-pyrrol-2-ylmethylene)hydrazide | |
>98% | |
Observed MW: 346.6 |
1378872-36-6 | |
C13H10Cl3N3O2 | |
25mg |
Safety and Handling
Recommended Storage : Store at 4°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title