ML 289, Tocris™

Selective negative allosteric modulator at mGlu3 receptors (IC50 = 660nM). Exhibits 15-fold selectivity for mGlu3 over mGlu2 Centrally penetrant

Manufacturer: R&D Systems 4976/50

Quantity 50mg

 View more versions of this product

Catalog No. 49-765-0

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1382481-79-9
    Molecular Formula C22H23NO3
    Formula Weight 349.42
    Synonym [(3R)-3-(Hydroxymethyl)-1-pipridinyl][4-[2-(4-methoxyphenyl)ethynyl]phenyl]methanone
    Chemical Name or Material ML 289

    Specifications

    Formula Weight 349.42
    Quantity 50mg
    Molecular Formula C22H23NO3
    CAS 1382481-79-9
    Synonym [(3R)-3-(Hydroxymethyl)-1-pipridinyl][4-[2-(4-methoxyphenyl)ethynyl]phenyl]methanone
    Recommended Storage Store at +4°C
    Chemical Name or Material ML 289

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit