Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 298 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
C22H23F3N4O2·HCl
3,4-Difluoro-N-[2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]dec-8-yl]ethyl]benzamide hydrochloride
This item is not returnable. View return policy
C22H23F3N4O2·HCl
3,4-Difluoro-N-[2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]dec-8-yl]ethyl]benzamide hydrochloride

Selective PLD2 inhibitor

Selective inhibitor of phospholipase D2 (PLD2) (IC50 values are 355 and >20,000 nM for PLD2 and PLD1, respectively). Decreases invasive migration in U87-MG glioblastoma cells, without affecting cell viability.
Quantity 50mg
Formula Weight 468.9
Percent Purity >98%
Chemical Name or Material ML 298 hydrochloride

Recommended Storage : Desiccate at Room Temperature