Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 315 hydrochloride Available on GSA/VA Contract for Federal Government customers only.

Low molecular weight, organic compound that may help regulate biological process.

$194.10 - $815.00

Specifications

CAS 1440251-53-5
Formula Weight 410.68g/mol
For Use With (Application) Cdc2-like Kinase Inhibitors
Molecular Formula C18H13Cl2N3O2.HCl
Storage Requirements Store at −20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-001-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5700/10
10mg Each for $194.10
Add to cart
 
57-005-0 View Documents Tocris Bioscience™
5700/50
50mg Each for $815.00
Add to cart
 
Description

Description

Inhibitor of cdc2-like kinases (Clk) and DYRK kinases (IC50 values are 68, 68 and 231 nM for Clk1, Clk4 and Clk2, respectively, and 282 and 1156 nM for Dyrk1A and Dyrk1B). Shows 20-fold selectivity for Clk4 over Dyrk1A. Stable in both mouse and human plasma. Moderately cell permeable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

1440251-53-5
Cdc2-like Kinase Inhibitors
Store at −20°C
410.68g/mol
C18H13Cl2N3O2.HCl
Cdc2-like Kinase

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit