Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ML 3403, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
549505-65-9
C23H21FN4S
404.507
VXPWQNBKEIVYIS-UHFFFAOYSA-N
p38 map kinase inhibitor iii,4-4-4-fluorophenyl-2-methylthio-1h-imidazol-5-yl-n-1-phenylethyl pyridin-2-amine,insolution p38 map kinase inhibitor iii,rs-4-5-4-fluorophenyl-2-methylsulfanyl-3h-imidazol-4-yl pyridin-2-yl-1-phenylethyl amine,d0w4ff,4-5-4-fluoro-phenyl-2-methylsulfanyl-3h-imidazol-4-yl-pyridin-2-yl-1-phenyl-ethyl-amine,5-4-4-fluorophenyl-2-methylthio-1h-imidazol-4-yl-n-1-phenylethyl-2-pyridinamine,4-5-4-fluorophenyl-2-methylsulfanyl-1h-imidazol-4-yl-n-1-phenylethyl pyridin-2-amine,2-pyridinamine, 4-4-4-fluorophenyl-2-methylthio-1h-imidazol-5-yl-n-1-phenylethyl,2-pyridinamine, 4-5-4-fluorophenyl-2-methylthio-1h-imidazol-4-yl-n-1-phenylethyl
6419739
4-[5-(4-fluorophenyl)-2-methylsulfanyl-1H-imidazol-4-yl]-N-(1-phenylethyl)pyridin-2-amine
CC(C1=CC=CC=C1)NC2=NC=CC(=C2)C3=C(NC(=N3)SC)C4=CC=C(C=C4)F
This item is not returnable. View return policy
549505-65-9
C23H21FN4S
404.507
VXPWQNBKEIVYIS-UHFFFAOYSA-N
p38 map kinase inhibitor iii,4-4-4-fluorophenyl-2-methylthio-1h-imidazol-5-yl-n-1-phenylethyl pyridin-2-amine,insolution p38 map kinase inhibitor iii,rs-4-5-4-fluorophenyl-2-methylsulfanyl-3h-imidazol-4-yl pyridin-2-yl-1-phenylethyl amine,d0w4ff,4-5-4-fluoro-phenyl-2-methylsulfanyl-3h-imidazol-4-yl-pyridin-2-yl-1-phenyl-ethyl-amine,5-4-4-fluorophenyl-2-methylthio-1h-imidazol-4-yl-n-1-phenylethyl-2-pyridinamine,4-5-4-fluorophenyl-2-methylsulfanyl-1h-imidazol-4-yl-n-1-phenylethyl pyridin-2-amine,2-pyridinamine, 4-4-4-fluorophenyl-2-methylthio-1h-imidazol-5-yl-n-1-phenylethyl,2-pyridinamine, 4-5-4-fluorophenyl-2-methylthio-1h-imidazol-4-yl-n-1-phenylethyl
6419739
4-[5-(4-fluorophenyl)-2-methylsulfanyl-1H-imidazol-4-yl]-N-(1-phenylethyl)pyridin-2-amine
CC(C1=CC=CC=C1)NC2=NC=CC(=C2)C3=C(NC(=N3)SC)C4=CC=C(C=C4)F

p38 inhibitor

p38 MAPK inhibitor (IC50 = 0.38 μM). Inhibits the release of IL-1β and TNF-α in a peripheral blood mononuclear cell (PBMC) assay (IC50 values are 0.039 and 0.16 μM respectively).
Quantity 50mg
Formula Weight 404.5
Percent Purity >98%
Chemical Name or Material ML 3403

Recommended Storage : Store at +4°C