ML 3403, Tocris™

p38 inhibitor

Manufacturer: R&D Systems 4586/10

Quantity 10mg

 View more versions of this product

Catalog No. 45-861-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C23H21FN4S
    Formula Weight 404.5
    Synonym [5-[4-(4-Fluorophenyl)-2-methylthio-1H-imidazol-4-yl]-N-(1-phenylethyl)-2-pyridinamine
    Chemical Name or Material ML 3403

    Specifications

    Formula Weight 404.5
    Quantity 10mg
    Assay Percent Range >98%
    Molecular Formula C23H21FN4S
    CAS 549505-65-9
    Synonym [5-[4-(4-Fluorophenyl)-2-methylthio-1H-imidazol-4-yl]-N-(1-phenylethyl)-2-pyridinamine
    Recommended Storage Store at +4°C
    Chemical Name or Material ML 3403

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit