Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ML 347
Potent and selective ALK1 and ALK2 inhibitor
$278.40 - $1141.44
Specifications
Purity | >98% |
---|---|
Gene Alias | LDN 193719, 1 lWY, VU0469381 |
Product Type | ML 347 |
Molecular Weight (g/mol) | 352.39 |
Inhibitors | ML 347 |
Description
Potent and selective ALK2 and ALK1 inhibitor (IC50 values are 32 and 46 nM, respectively); displays >200 fold selectivity over ALK3 and ALK6 and >400 fold selectivity over VEGF2. Exhibits no activity at ALK4, ALK5 or in a panel of related kinases. Inhibits BMP4 signaling in a functional assay (IC50 = 152 nM).Specifications
>98% | |
ML 347 | |
ML 347 |
LDN 193719, 1 lWY, VU0469381 | |
352.39 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title