Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 382 Available on GSA/VA Contract for Federal Government customers only.

MrgX1 (SNSR4) positive allosteric modulator

$146.90 - $606.00

Specifications

Chemical Name or Material Mas-Related G Protein Coupled Receptor Modulators
Molecular Formula C18H20N2O4S
CAS 1646499-97-9
Product Type Target: Sensory Neuron-Specific Receptor Agonists
Formula Weight 360.43
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-381-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5738/10
10mg Each for $146.90
Add to cart
 
57-385-0 View Documents Tocris Bioscience™
5738/50
Each for $606.00
Add to cart
 
Description

Description

Selective MRGPRX1 positive allosteric modulator (EC50 = 190 nM). Exhibits selectivity for MrgX1 over MrgX2, MrgC11 and a panel of 68 other GPCRs, ion channels and transporters. Enhances BAM (8-22) potency at MRGPRX1 by 7-fold.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Mas-Related G Protein Coupled Receptor Modulators
1646499-97-9
360.43
C18H20N2O4S
Target: Sensory Neuron-Specific Receptor Agonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at RT

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit