Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ ML 402

Selective TREK channel activator

$215.76 - $902.48

Specifications

Chemical Name or Material N-[2-(4-Chloro-2-methylphenoxy)ethyl]-2-thiophenecarboxamide
CAS 298684-44-3
Molecular Formula C14H14ClNO2S
Purity 98%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
68-881-0
View Documents
Tocris™
6888/10
10 mg
Each for $215.76
Add to Cart
 
68-885-0
View Documents
Tocris™
6888/50
50 mg
Each for $902.48
Add to Cart
 
Description

Description

Selectively activates K2P2.1 (TREK-1) and K2P10.1 (TREK-2) with no significant effect at K2P4.1 channels (TRAAK).
Specifications

Specifications

N-[2-(4-Chloro-2-methylphenoxy)ethyl]-2-thiophenecarboxamide
298684-44-3
C14H14ClNO2S
98%
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit