Learn More
Medchemexpress LLC ML00253764 | 681847-92-7 | 99.7% | 377.25 g/mol | C18H18BrFN2O | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12474010MM 1ML
Encompass_Preferred
ML00253764 is a selective melanocortin receptor 4 (MC4R) antagonist reported to induce apoptosis by inhibiting ERK1/2 and Akt phosphorylation and to have anticancer activity. It is supplied as a solid (white to off-white) and as a 10 mM solution in DMSO, with high purity and recommended storage conditions for powder and solvent.
  • Selective MC4R antagonist with reported anticancer activity.
  • Induces apoptosis via inhibition of ERK1/2 and Akt phosphorylation.
  • High purity (99.7%) suitable for research applications.
  • Available as solid and as 10 mM solution in DMSO.
  • Multiple solid pack sizes (5-100 mg) for experimental flexibility.
  • Recommended storage conditions provided to preserve stability.
Catalog No. 50-002-35879