Learn More
Selleck Chemical LLCSupplier Diversity Partner ML130 (Nodinitib-1) DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S28635MG
ML130 (Nodinitib-1) is a potent and selective inhibitor of NOD1 with IC50 of 0.56 μM, inhibits NF-κB activation, exhibits 36-fold selectivity over NOD2.
Catalog No. 50-194-8078