Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLC1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537933
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
MLC1 may be a transporter. It may act as a non-selective neuronal cation channel.The function of this gene product is unknown; however, homology to other proteins suggests that it may be an integral membrane transporter. Mutations in this gene have been associated with megalencephalic leukoencephalopathy with subcortical cysts, an autosomal recessive neurological disorder. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
MLC1 | |
Polyclonal | |
Unconjugated | |
MLC1 | |
AW048630; BB074274; KIAA0027; Kiaa0027-hp; LVM; megalencephalic leukoencephalopathy with subcortical cysts 1; megalencephalic leukoencephalopathy with subcortical cysts 1 gene product; megalencephalic leukoencephalopathy with subcortical cysts 1 homolog; megalencephalic leukoencephalopathy with subcortical cysts 1 homolog (human); membrane protein MLC1; mKIAA0027; MLC; MLC1; modulator of VRAC current 1; VL; WKL1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
23209 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q15049 | |
MLC1 | |
Peptide with sequence C-SDSANILDEVPFPAR, from the internal region of MLC1 (aa179-193). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction