Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLF1 Mouse anti-Human, Clone: OTI5D9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504745

Catalog No. 50-167-5365


Add to cart

Description

Description

MLF1 is involved in lineage commitment of primary hemopoietic progenitors by restricting erythroid formation and enhancing myeloid formation. The protein interferes with erythopoietin-induced erythroid terminal differentiation by preventing cells from exiting the cell cycle through suppression of CDKN1B/p27Kip1 levels. It suppresses RFWD2/COP1 activity via CSN3 which activates p53 and induces cell cycle arrest. It binds DNA and affects the expression of a number of genes so may function as a transcription factor in the nucleus.
Specifications

Specifications

MLF1
Monoclonal
1.22 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P58340
myeloid leukemia factor 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4291
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI5D9
Unconjugated
MLF1
Liquid
MLF1
Full length human recombit protein of human MLF1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit