Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLF2 Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: OTI1E3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272721C
Description
MLF2 Monoclonal specifically detects MLF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MLF2 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MLF2 | |
Full length recombinant protein of human MLF2 (NP_005430) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E3 | |
Western Blot, Immunohistochemistry | |
Myelodysplasia-myeloid leukemia factor 2, myeloid leukemia factor 2, NTN4 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
8079.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title