Learn More
Aviva Systems Biology Corp MLH1 Antibody - N-terminal region (ARP56076_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56076_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MLH1. Alternate names: COCA2, FCC2, HNPCC, HNPCC2, MGC5172, hMLH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61710