Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLKL Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen MLKL
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093699
View Documents
Novus Biologicals
NBP303885100UL
100 μg Each for $320.71
Add to cart
 
NB093700
View Documents
Novus Biologicals
NBP30388520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

MLKL Polyclonal antibody specifically detects MLKL in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

MLKL
Unconjugated
Rabbit
Necroptosis, Protein Kinase
PBS with 50% glycerol, pH7.3.
197259
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, Western Blot
Purified
RUO
Human, Mouse
FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein
Recombinant protein of mouse MLKL.
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit