Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103371
Description
Antibody detects endogenous levels of total WBP7.
This gene encodes a protein which contains multiple domains including a CXXC zinc finger, three PHD zinc fingers, two FY-rich domains, and a SET domain. The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL family. This gene is ubiquitously expressed in adult tissues. It is also amplified in solid tumor cell lines, and may be involved in human cancer. Two alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene, however, the full length nature of the shorter transcript is not known.
Specifications
MLL4 | |
Polyclonal | |
Unconjugated | |
KMT2B | |
2610014H22Rik; ALL1-related protein; ALR; BC032281; BC058659; C430014K11Rik; CXXC10; histone-lysine N-methyltransferase 2B; histone-lysine N-methyltransferase 2B-like; histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; histone-lysine N-methyltransferase MLL4; HRX2; KIAA0304; Kmt2b; Kmt2d; LOC102550344; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase 2B; lysine (K)-specific methyltransferase 2B; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2B; lysine N-methyltransferase 2B; lysine N-methyltransferase 2D; mixed lineage leukemia 2; mixed lineage leukemia gene homolog 2; mKIAA0304; MLL1B; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila) 4; myeloid/lymphoid or mixed-lineage leukemia 2; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia protein 2; myeloid/lymphoid or mixed-lineage leukemia protein 4; Myeloid/lymphoid or mixed-lineage leukemia protein 4 homolog; trithorax homolog 2; trithorax homologue 2; TRX2; Wbp7; WBP-7; WW domain binding protein 7; WW domain-binding protein 7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
102550344, 75410, 9757 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O08550, Q9UMN6 | |
KMT2B | |
A synthesized peptide derived from human KMT2B(Accession Q9UMN6), corresponding to amino acid residues T2687-N2715. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction